missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3PXD2A Monoclonal Antibody (OTI1F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811757
This item is not returnable.
View return policy
Description
Adapter protein involved in invadopodia and podosome formation, extracellular matrix degradation and invasiveness of some cancer cells. Binds matrix metalloproteinases (ADAMs), NADPH oxidases (NOXs) and phosphoinositides. Acts as an organizer protein that allows NOX1- or NOX3-dependent reactive oxygen species (ROS) generation and ROS localization. In association with ADAM12, mediates the neurotoxic effect of amyloid-beta peptide.Specifications
| SH3PXD2A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5TCZ1 | |
| SH3PXD2A | |
| Human recombit protein fragment corresponding to amino acids 299-395 of human SH3PXD2A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1F5 | |
| Unconjugated | |
| SH3PXD2A | |
| FISH, SH3MD1, TKS5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9644 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction