missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3PXD2A Monoclonal Antibody (OTI1F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811757S
This item is not returnable.
View return policy
Description
SH3PXD2A Monoclonal antibody specifically detects SH3PXD2A in Human samples. It is validated for Western BlotSpecifications
| SH3PXD2A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P32322 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5831 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1F5 | |
| Unconjugated | |
| PYCR1 | |
| 2310014D11Rik; AA589508; Adapter protein TKS5; adaptor protein TKS5; AI256723; AI413738; C230050L11; EG329070; FISH; Five SH3 domain-containing protein; five SH3 domains; Gm5098; KIAA0418; SH3 and PX domain-containing protein 2A; SH3 and PX domains 2A; SH3 multiple domains 1; SH3 multiple domains protein 1; Sh3md1; SH3PXD2A; Tks5; TSK5; Tyrosine kinase substrate with five SH3 domains | |
| Human recombinant protein fragment corresponding to amino acids 1-319 of human PYCR1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction