missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Activates both Ras-dependent and Ras-independent migratory pathways in melanomas. Contributes to the early phases of agrin-induced tyrosine phosphorylation of CHRNB1.
Specifications
Specifications
| Antigen | SHC4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SHC4 |
| Gene Accession No. | Q6S5L8, Q6S5L9 |
| Gene Alias | hShcD, Rai like protein, RaLP, SH2 domain protein C4, SHC transforming protein 4, SHC transforming protein D, SHC4, SHCD |
| Gene Symbols | SHC4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?