missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Canine IgG Fc Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP2-69475
352.75 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Canine samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoelectrophoresis
Specifications
| IgG Fc | |
| Polyclonal | |
| Unconjugated | |
| Whole antiserum | |
| Highly purified normal IgG isolated from pooled dog serum was used for immunization of the sheep. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA, Immunofluorescence, Immunoelectrophoresis | |
| LYOPH | |
| ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoelectrophoresis | |
| Sheep | |
| Whole Antiserum | |
| RUO | |
| Secondary | |
| Canine | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction