Learn More
Sheep anti-Guinea Pig IgM Fc Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69694
352.75 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18160423
  • 481.00 € / 1mL
Description
IgM Fc Polyclonal antibody specifically detects IgM Fc in Guinea Pig samples. It is validated for Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgM Fc
Polyclonal
Unconjugated
Whole antiserum with No Preservative
Highly purified normal IgM isolated from pooled guinea pig serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
The reactivity of the antiserum is restricted to the Fc part of the IgM molecule. In immunoelectrophoresis and radial immunodiffusion, using various antiserum concentrations against normal guinea pig serum a single precipitin line is obtained which shows a reaction of identity with the precipitin line obtained with purified IgM. No precipitation reaction is obtained with purified IgG, IgA, and IgG/Fab fragments.
Guinea Pig
Lyophilized
Immunoprecipitation
LYOPH
Immunoprecipitation 1:10 - 1:500
Sheep
Whole Antiserum
Secondary
Reconstitute the lyophilized antiserum by adding 1 ml sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.