Learn More
Sheep anti-Human IgA/Secretory Component Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69294
Product Code. 18603307
  • 443.00 € / 1mL
Description
IgA/Secretory Component Polyclonal antibody specifically detects IgA/Secretory Component in Human samples. It is validated for Immunoelectrophoresis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgA/Secretory Component
Polyclonal
Unconjugated
Sheep
Whole Antiserum
RUO
Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunoelectrophoresis
LYOPH
Whole antiserum
Highly purified IgA/Secretory Component isolated from pooled human milk. Freund's complete adjuvant is used in the first step of the immunization procedure.
1 mL
Secondary
Human
Lyophilized