missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Human IgD Fc Secondary Antibody, Novus Biologicals™

Product Code. 18677376 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18677376 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18677376 Supplier Novus Biologicals Supplier No. NBP269331

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IgD Fc Polyclonal antibody specifically detects IgD Fc in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoelectrophoresis
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgD Fc
Applications Immunohistochemistry (Paraffin), Immunoelectrophoresis
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:10 - 1:500, Immunoelectrophoresis
Formulation Delipidated, heat inactivated, lyophilized, stable whole antiserum
Host Species Sheep
Immunogen Pools of purified homogenous IgD isolated from human serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Whole Antiserum
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Reconstitution Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Lyophilized
Show More Show Less
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.