missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHPRH Monoclonal Antibody (OTI6D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501465
This item is not returnable.
View return policy
Description
A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus suggests a role in tumor suppression.Specifications
| SHPRH | |
| Monoclonal | |
| 0.97 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q149N8 | |
| SHPRH | |
| Human recombit protein fragment corresponding to amino acids 1-100 and 1500-1600 of human SHPRH produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI6D9 | |
| Unconjugated | |
| SHPRH | |
| bA545I5.2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 257218 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction