missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SHROOM4 Polyclonal Antibody

Product Code. 17874250
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17874250 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17874250 Supplier Invitrogen™ Supplier No. PA5139736
  • ONLINE EXCLUSIVE PRICE
    464.00 €
    329.44 € / 100µL
    Save 134.56 €
    29% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GSCALNPEEV LEQPQPLSFG HLEGSRQGSQ SVPAEQESFA LHSSDFLPPI RGHLGSQPEQ AQPPCYYGIG GLWRTSGQEA TESAKQEFQH FSPPSGAPGI PTSYSAYYNI SVAKAELLNK LKDQPEMAEI G Highest antigen sequence indentity to the following orthologs: Mouse - 73%, Rat - 69%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SHROOM4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SHROOM4
Gene Accession No. Q9ULL8
Gene Alias KIAA1202; Protein Shroom4; second homolog of apical protein; SHAP; shroom family member 4; SHROOM4
Gene Symbols SHROOM4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SHROOM4. Recombinant protein control fragment (Product #RP-92222).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57477
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.