missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIAE Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93817-0.02ml
This item is not returnable.
View return policy
Description
SIAE Polyclonal antibody specifically detects SIAE in Human samples. It is validated for Western Blot
Specifications
| SIAE | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| AIS6, CSEC, CSE-C, cytosolic sialic acid 9-O-acetylesterase homolog, EC 3.1.1.53, H-Lse, LSE, MGC87009, sialate O-acetylesterase, sialic acid acetylesterase, Sialic acid-specific 9-O-acetylesterase, sialic acid-specific acetylesterase II, YSG2, Ysg2 homolog (mouse) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 270-450 of human SIAE (NP_733746.1). LKGVVWYQGESNINYNTDLYNCTFPALIEDWRETFHRGSQGQTERFFPFGLVQLSSDLSKKSSDDGFPQIRWHQTADFGYVPNPKMPNTFMAVAMDLCDRDSPFGSIHPRDKQTVAYRLHLGARALAYGEKNLTFEGPLPEKIELLAHKGLLNLTYYQQIQVQKKDNKIFEISCCSDHRCK | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54414 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction