missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIAH2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 468.00 €
Specifications
| Antigen | SIAH2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SIAH2 Polyclonal antibody specifically detects SIAH2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| SIAH2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 6478 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| E3 ubiquitin-protein ligase SIAH2, EC 6.3.2, EC 6.3.2.-, hSiah2, seven in absentia (Drosophila) homolog 2, Seven in absentia homolog 2, seven in absentia homolog 2 (Drosophila), siah-2 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human SIAH2 (NP_005058.3). VCNQCRQKLSCCPTCRGALTPSIRNLAMEKVASAVLFPCKYATTGCSLTLHHTEKPEHEDICEYRPYSCPCPGASCKWQGSLEAVMSHLMHAHKSITTLQG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title