missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIAH2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93257-0.02ml
This item is not returnable.
View return policy
Description
SIAH2 Polyclonal antibody specifically detects SIAH2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SIAH2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| E3 ubiquitin-protein ligase SIAH2, EC 6.3.2, EC 6.3.2.-, hSiah2, seven in absentia (Drosophila) homolog 2, Seven in absentia homolog 2, seven in absentia homolog 2 (Drosophila), siah-2 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human SIAH2 (NP_005058.3). VCNQCRQKLSCCPTCRGALTPSIRNLAMEKVASAVLFPCKYATTGCSLTLHHTEKPEHEDICEYRPYSCPCPGASCKWQGSLEAVMSHLMHAHKSITTLQG | |
| 0.02 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication | |
| 6478 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction