missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sialin/SLC17A5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94758-0.02ml
This item is not returnable.
View return policy
Description
Sialin/SLC17A5 Polyclonal antibody specifically detects Sialin/SLC17A5 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| Sialin/SLC17A5 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| ASTSodium/sialic acid cotransporter, FLJ22227, FLJ23268, ISSD, Membrane glycoprotein HP59, NSD, SD, sialic acid storage disease, SIALIN, SIASD, SLDSolute carrier family 17 member 5, solute carrier family 17 (anion/sugar transporter), member 5, solute carrier family 17, member 5 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC17A5 (NP_036566.1). MRSPVRDLARNDGEESTDRTPLLPGAPRAEAAPVCCSARYNLAILAFFGFFIVYALRVNLSVALVDMVDSNTTLEDNRTSKACPEHSAPIKVHHNQTGKK | |
| 0.02 mL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26503 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction