missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Sierra™ Sterile Facemasks

Product Code. 15684894 Shop All DuPont Products
Change view
Click to view available options
Width (Metric): 23 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15684894 Width (Metric): 23 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15684894 Supplier DuPont™ Supplier No. D13473980

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides process protection from human contamination. Dupont™ Sierra™ Sterile Facemasks include bound ties and pleated rayon outer facing. Gamma sterilized.

  • Gamma-sterilized
  • Pleated rayon outer facing
  • Bound Tyvek™ ties
  • Metal nose piece.
  • White, one size fits most
  • 23 cm width
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Face Mask
Color White
Material Rayon
Size Standard
Sterility Sterile
Certifications/Compliance Sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1)
Certifications/Compliance (Europe) Suitable for use in GMP class C/D cleanrooms
Cleanroom Class Class 1000 (ISO 6)
Disposable Single-use
For Use With (Application) Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Controlled Environments
Includes Bound Ties, Metal Nose Piece
Model D13473980
Width (Metric) 23 cm
Show More Show Less

The user should make the risk assessment to determine the protection required.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.