Learn More
Endecotts Sieve Brush Double Ended Nylon, One Hard and One Soft End
Product Code. 12904175
Change view
Click to view available options
For Use With (Equipment):
For use with Sieves
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12904175 For use with Sieves Each
1 options
Product Code. 12904175 Supplier Endecotts Supplier No. ZSXSBDEND2
Only null left
Add to basket
Add to basket
Large double-ended nylon sieve brush with interchangeable hard and soft ends.
  • Large double-ended sieve brush made from durable nylon, designed with one hard end and one soft end to support effective and controlled cleaning of test sieves without damaging the mesh.
Specifications
For Use With (Equipment) For use with Sieves