Learn More
Invitrogen™ Sifalimumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558635
 View more versions of this product
Product Code. 30284589
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Sifalimumab is a fully human monoclonal antibody targeting interferon-alpha. The levels of interferon-alpha are elevated in many patients with active systemic lupus erythematosus (SLE, or lupus) and other autoimmune disorders, and may be associated with disease activity. Sifalimumab may suppress the abnormal immune activity associated with lupus by binding to multiple interferon-alpha subtypes seen in the serum of lupus patients.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sifalimumab
Recombinant Monoclonal
Unconjugated
MDX-1103; MEDI-545
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized