missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-6 (CD327) Mouse anti-Human, Brilliant Violet 786, Clone: 767329, BD OptiBuild™
Description
Siglec-6 (CD327) Monoclonal antibody specifically detects Siglec-6 (CD327) in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | Siglec-6 (CD327) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 767329 |
| Concentration | 0.2 mg/mL |
| Conjugate | Brilliant Violet 786 |
| Host Species | Mouse |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?