Learn More
Siglec-G Antibody (805903), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB7775Z
Product Code. 18549768
  • 664.00 € / 0.10mL
Description
Siglec-G Monoclonal antibody specifically detects Siglec-G in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Siglec-G
Monoclonal
DyLight 755
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
89790
Store at 4°C in the dark.
IgG2a
Flow Cytometry, CyTOF
805903
Flow Cytometry, CyTOF-ready
PRO940;Sialic acid-binding Ig-like lectin 10;SIGLEC10;SIGLEC-10;SLG2
Chinese hamster ovary cell line CHO-derived recombinant mouse Siglec-G, Glu20-Gln525, Accession # NP_766488
0.1 mL
Primary
Mouse
Purified