missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGNR1/CD209b Rabbit anti-Mouse, Clone: 123, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90608-100UL
This item is not returnable.
View return policy
Description
SIGNR1/CD209b Monoclonal specifically detects SIGNR1/CD209b in Mouse samples. It is validated for Sandwich ELISA Capture.
Specifications
| SIGNR1/CD209b | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 170786 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 123 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| CD209 antigen like protein B, DC SIGN related protein 1, DC SIGNR1, OtB7 | |
| Recombinant Mouse SIGNR1/CD209b Protein | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur