missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIK2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149056
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
SNF1LK2 (QIK) is a serine/threonine kinase and is a salt-inducible kinase that belongs to the AMP-activated protein kinase family. SNF1LK2 negatively regulates CRE-binding protein (CREB) activity by phosphorylating the CREB-specific coactivator transducer of regulated CREB activity (TORC). SIK2 is thought to be part of a signaling cascade that regulates the expression and activity of the insulin-induced genes PGC-1'alpha and UCP-1 in brown adipocytes, impairment of which has been implicated in obesity and insulin resistance in human and animal models. SIK2 has also been reported as a key regulator for neuronal survival after ischemia, suppressing CREB-mediated gene expression after oxygen-glucose deprivation.
Specifications
| SIK2 | |
| Polyclonal | |
| Unconjugated | |
| SIK2 | |
| G630080D20Rik; KIAA0781; LOH11CR1I; QIK; Qin-induced kinase; RGD1559698; salt induceable kinase 2; salt inducible kinase 2; salt-inducible kinase 2; salt-inducible protein kinase 2; salt-inducible serine/threonine kinase 2; serine/threonine-protein kinase SIK2; Serine/threonine-protein kinase SNF1-like kinase 2; Sik2; SIK-2; SNF1-like kinase 2; SNF1LK2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 23235 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9H0K1 | |
| SIK2 | |
| E. coli-derived recombinant human SNF1LK2, Ala663-Leu818. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction