missing translation for 'onlineSavingsMsg'
Learn More

SIK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

203.00 € - 589.00 €

Specifications

Antigen SIK3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18746153
missing translation for 'viewDocuments'
Novus Biologicals
NBP2-47278
0.1 mL
589.00 €
0.10mL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
18672938
missing translation for 'viewDocuments'
Novus Biologicals
NBP2-52411-0.025mg
25 μL
203.00 €
0.03mg
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
18660326
missing translation for 'viewDocuments'
Novus Biologicals
NBP2-47278-25ul
25 μL
415.00 €
25µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SIK3 Polyclonal specifically detects SIK3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

SIK3
Polyclonal
Rabbit
Protein Kinase
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
23387
This antibody was developed against a recombinant protein corresponding to amino acids: QQEYQELFRHMNQGDAGSLAPSLGGQSMTERQALSYQNADSYHHHTSPQHLLQIRAQECVSQASSPTPPHGYAHQPALMHSESMEEDCSCEGAKDGFQD
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse
EC 2.7.11, EC 2.7.11.1, KIAA0999L19, QSKFLJ12240, Salt-inducible kinase 3, Serine/threonine-protein kinase QSK, serine/threonine-protein kinase SIK3, SIK family kinase 3, SIK-3
SIK3
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.