missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Silencer™ Pre-Designed siRNA

Product Code. 10167104
Change view
Click to view available options
Product Type:
Silencer™ Pre-Designed HPLC siRNA
Silencer™ Pre-Designed In Vivo siRNA
Silencer™ Pre-Designed Standard (Made to Order) siRNA
Silencer™ Pre-Designed Standard siRNA
Silencer™ Pre-Designed siRNA
Purity:
HPL-OLIGOC
HPLC
In Vivo
Standard
Label or Dye:
CY™3
FAM
Unlabeled
Quantity:
1 μmol
10 μmol
20 nmol
250 nmol
40 nmol
5 nmol
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Purity Label or Dye Quantity Unit Size
10167104 Silencer™ Pre-Designed Standard siRNA Standard Unlabeled 20 nmol Each
10446914 Silencer™ Pre-Designed siRNA Standard Unlabeled 5 nmol Each
10289574 Silencer™ Pre-Designed Standard siRNA Standard Unlabeled 40 nmol Each
10566735 Silencer™ Pre-Designed Standard (Made to Order) siRNA Standard Unlabeled 5 nmol Each
10370495 Silencer™ Pre-Designed Standard (Made to Order) siRNA Standard Unlabeled 20 nmol Each
10015364 Silencer™ Pre-Designed Standard (Made to Order) siRNA Standard Unlabeled 40 nmol Each
10418844 Silencer™ Pre-Designed HPLC siRNA HPL-OLIGOC Unlabeled 250 nmol Each
10301135 Silencer™ Pre-Designed In Vivo siRNA In Vivo Unlabeled 1 μmol Each
10497104 Silencer™ Pre-Designed In Vivo siRNA In Vivo Unlabeled 10 μmol Each
10456914 Silencer™ Pre-Designed siRNA HPLC CY™3 20 nmol Each
10341085 Silencer™ Pre-Designed siRNA HPLC FAM 20 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 10167104 Supplier Ambion™ Supplier No. AM16704
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.

Silencer Predesigned siRNAs are high-quality, synthetic small interfering RNAs (siRNAs) designed for gene silencing applications in mammalian cells. This cost-effective RNA interference tool delivers reliable gene knockdown performance while maintaining economic accessibility for research applications. Our siRNA products are meticulously crafted to help ensure high efficiency and specificity, enabling researchers to achieve reliable and reproducible gene knockdown results. Silencer Predesigned siRNAs are unmodified synthetic RNA duplexes with a 19-mer duplex and two-nucleotide 3' overhangs.

Key features include:

  • High silencing efficiency—demonstrates good knockdown performance with optimized target recognition for effective gene expression suppression
  • Enhanced specificity—algorithm designed for precise gene targeting to minimize off-target effects and help ensure selective gene silencing
  • Guaranteed performance—two out of three predesigned siRNAs will result in >70% gene knockdown. Validated designs available for many common genes.
  • Cost-effective solution—exceptional value for research applications without compromising performance quality

Streamlined workflow for rapid results

Utilizing our gene knockdown technology is straightforward and efficient. Simply order siRNAs for the desired gene targets, deliver to the cells, and detect knockdown efficiency and validate the resulting phenotype. Take advantage of our validated designs for better confidence in your experiments. This streamlined process allows for rapid experimental progress and accelerated discovery in gene function studies.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Silencer™ Select siRNAs are the result of recent improvements in siRNA design combined with novel chemical modifications yielding unrivaled knockdown efficiency and specificity. Our best performing siRNA for cell culture work. Silencer™ siRNAs are unmodified 21 nucleotide siRNAs that have been designed with an older, yet rigorously tested design algorithm. A lower cost alternative to Silencer™ Select siRNAs, but without some of the performance enhancements.
Description Silencer Pre-Designed siRNA STD, 20nmol, Incorporate the Latest Improvements in siRNA Design, Off-Target Effect Prediction Algorithms, and Chemistry, Guaranteed-to-Silence Based on their Proven Design
Format 96-well Plate, 384-well Plate
RNAi Type siRNA
Sample Type mRNA
Final Product Type Silencer Pre-Designed siRNA STD
Label or Dye Unlabeled
Product Line Silencer, Ambion
Product Type Silencer™ Pre-Designed Standard siRNA
Purity Standard
Quantity 20 nmol
Show More Show Less
Where should I add a fluorescent tag to minimize any negative functional effect on my siRNA?

For any RNAi duplex (miRNA or siRNA) there is a possibility that adding a modification will affect how the antisense strand interacts with RISC and aligns with its mRNA target, thus impacting functionality. For that reason, we recommend that you label the 5' end of the sense strand for siRNA (or passenger strand for miRNA). Since the 5' end of the antisense (guide strand) is what gets recognized by RISC, it is best to add the fluorescent modification to the end of the duplex on the strand that does not interact with RISC. You can also test a labeled vs. non-labeled RNAi duplex to determine if the modification is affecting the siRNA or miRNA functionality.

How can I find the GC content for the predesigned Silencer siRNA or Silencer Select siRNA sequences prior to purchase?

Please contact Technical Support at techsupport@thermofisher.com to request the GC content for one of our predesigned Silencer siRNA or Silencer Select siRNA sequences.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.