missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Silencer™ Validated siRNA

Product Code. 10734237
Change view
Click to view available options
Purity:
CY3 Labeled
FAM Labeled
HPL-OLIGOC
HPLC
In Vivo
Standard
Quantity:
1 μmol
10 μmol
20 nmol
250 nmol
40 nmol
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Purity Quantity Unit Size
10734237 FAM Labeled 20 nmol Each
10534075 Standard 40 nmol Each
10724047 HPLC 40 nmol Each
10177964 HPL-OLIGOC 250 nmol Each
10515005 In Vivo 1 μmol Each
10085064 In Vivo 10 μmol Each
10289494 In Vivo 250 nmol Each
10724237 CY3 Labeled 20 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10734237 Supplier Ambion™ Supplier No. AM51327
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.

  • The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry
  • Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression
  • Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening
  • The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment

The benefits of using Silencer™ Select siRNAs include:

  • Potent – up to 100-fold more potent than currently available siRNAs and fewer off-target effects
  • Most Specific – LNA™ chemical modifications reduce off-target effects by up to 90%
  • Reliable – demonstrated improvements in consistency and reliability of phenotypic results
  • Guaranteed – 100% guaranteed to silence, the best guarantee in the industry

Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.

Approximate Turnaround Times*

  • Standard purification: 4 business days
  • HPLC purification: 6 business days (20 and 40 nmol); 15 business days (250 nmol)
  • In Vivo Ready: 15 business days (250 nmol)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Silencer™ Validated siRNA
RNAi Type siRNA
Sample Type RNA
Product Line Silencer
Purity FAM Labeled
Quantity 20 nmol
Temperature (Metric) Store at room temperature
Where should I add a fluorescent tag to minimize any negative functional effect on my siRNA?

For any RNAi duplex (miRNA or siRNA) there is a possibility that adding a modification will affect how the antisense strand interacts with RISC and aligns with its mRNA target, thus impacting functionality. For that reason, we recommend that you label the 5' end of the sense strand for siRNA (or passenger strand for miRNA). Since the 5' end of the antisense (guide strand) is what gets recognized by RISC, it is best to add the fluorescent modification to the end of the duplex on the strand that does not interact with RISC. You can also test a labeled vs. non-labeled RNAi duplex to determine if the modification is affecting the siRNA or miRNA functionality.

How can I find the GC content for the predesigned Silencer siRNA or Silencer Select siRNA sequences prior to purchase?

Please contact Technical Support at techsupport@thermofisher.com to request the GC content for one of our predesigned Silencer siRNA or Silencer Select siRNA sequences.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.