missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Silencer™ Select Pre-Designed siRNA

Product Code. 10574835
Change view
Click to view available options
Description:
Silencer™ Select Validated siRNA, 20nmol, HPLC Purity, –20°C Storage, Potent, Most Specific, Reliable, Guaranteed, Incorporate the Latest Improvements in siRNA design, Off-Target Effect Prediction Algorithms and Chemistry
Silencer™ Select Validated siRNA, 40nmol, HPLC Purity, –20°C Storage, Potent, Most Specific, Reliable, Guaranteed, Incorporate the Latest Improvements in siRNA design, Off-Target Effect Prediction Algorithms and Chemistry
Silencer™ Select Validated, 250nmol, HPLC Purity, –20°C Storage, Classic 21-mers which Incorporate the Latest Improvements in siRNA Design, Off-target Effect Prediction Algorithms and Chemistry, Silencer Select Pre-Designed siRNAs are Guaranteed-To-Silence Based on their Proven Design
Quantity:
20 nmol
250 nmol
40 nmol
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Unit Size
10574835 Silencer™ Select Validated, 250nmol, HPLC Purity, –20°C Storage, Classic 21-mers which Incorporate the Latest Improvements in siRNA Design, Off-target Effect Prediction Algorithms and Chemistry, Silencer Select Pre-Designed siRNAs are Guaranteed-To-Silence Based on their Proven Design 250 nmol Each
10157534 Silencer™ Select Validated siRNA, 20nmol, HPLC Purity, –20°C Storage, Potent, Most Specific, Reliable, Guaranteed, Incorporate the Latest Improvements in siRNA design, Off-Target Effect Prediction Algorithms and Chemistry 20 nmol Each
10299774 Silencer™ Select Validated siRNA, 40nmol, HPLC Purity, –20°C Storage, Potent, Most Specific, Reliable, Guaranteed, Incorporate the Latest Improvements in siRNA design, Off-Target Effect Prediction Algorithms and Chemistry 40 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10574835 Supplier Ambion™ Supplier No. 4407270
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Silencer™ Select Pre-Designed siRNA

The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry. A choice of Silencer™ Select siRNAs are available—Pre-Designed or Validated. Silencer™ Select Pre-Designed siRNAs are guaranteed-to-silence based on their proven design while the Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression. Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening. The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment.

The benefits of using Silencer™ Select siRNAs include:
  • Potent—up to 100-fold more potent than currently available siRNAs and fewer off-target effects
  • Most Specific—LNA™ chemical modifications reduce off-target effects by up to 90%
  • Reliable—demonstrated improvements in consistency and reliability of phenotypic results
  • Guaranteed—100% guaranteed to silence, the best guarantee in the industry

  • Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.

    Approximate Turnaround Times*
  • Standard purification: 4 business days
  • HPLC purification: 6 business days (20 and 40 nmol); 15 business days (250 nmol)
  • In Vivo Ready: 15 business days (250 nmol)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage -20°C
    Description Silencer™ Select Validated, 250nmol, HPLC Purity, –20°C Storage, Classic 21-mers which Incorporate the Latest Improvements in siRNA Design, Off-target Effect Prediction Algorithms and Chemistry, Silencer Select Pre-Designed siRNAs are Guaranteed-To-Silence Based on their Proven Design
    Product Type Silencer™ Select Validated
    Purity HPLC
    Quantity 250 nmol

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.