missing translation for 'onlineSavingsMsg'
Learn More

Silicon for AAS 1g/L H2O matrix, Fisher chemical™
BIOPHARMA

Product Code. 12973215 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Packaging:
Bottle
Unit Size:
1000mL
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12973215 100 mL Bottle 100mL
12993215 500 mL Bottle 500mL
12983215 1000 mL Bottle 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12973215 Supplier Fisher Chemical Supplier No. B 053 100
  • ONLINE EXCLUSIVE PRICE
    27.00 €
    25.92 € / 100mL
    Save 1.08 €
    4% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula Si

Specifications

Packaging Bottle
Physical Form Liquid
Concentration 1 g/L ±0.002 g/L
Type Flame Single-element Standard Solution
Solvent or Matrix Water
Quantity 100 mL
Chemical Name or Material Silicon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.