missing translation for 'onlineSavingsMsg'
Learn More

Silicon Standard Metal Solution 1000ppm, for AAS, Solution Aqueous, Fisher Chemical™

Product Code. 10214062 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10214062 100 mL 100mL
10030220 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10214062 Supplier Fisher Chemical Supplier No. J/8062/05
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Silicon Standard Metal Solution 1000ppm, for AAS, Solution Aqueous

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16919-19-0
Molecular Formula F6H8N2Si
Molecular Weight (g/mol) 178.153
MDL Number 85311
InChI Key ITHIMUMYFVCXSL-UHFFFAOYSA-P
PubChem CID 28145
IUPAC Name diazanium;hexafluorosilicon(2-)
SMILES [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F

Specifications

Packaging HDPE plastic bottle
Physical Form Liquid
Color Colorless
Quantity 100 mL
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.