missing translation for 'onlineSavingsMsg'
Learn More

Silodosin, MedChemExpress

Product Code. 16434556
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16434556 100 mg 100mg
16433308 10 mg 10mg
16419846 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16434556 Supplier Medchem Express Supplier No. HY10122
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Silodosin (KAD 3213; KMD 3213) is a potent, selective and orally active α1A-adrenergic receptor (α1A-AR) blocker. Silodosin exhibits high affinity for α1A-AR (Ki=0.036 nM), over 162-fold and 50-fold than for α1B-AR and α1D-AR with Ki values of 21 nM and 2.0 nM, respectively. Silodosin is an effective and well-tolerated agent, it can be used for the investigation of LUTS/BPH.

Silodosin (KAD 3213

Specifications

Chemical Name or Material Silodosin
CAS 160970-54-7
Color White
Purity Grade Notes Research
Molecular Formula C25H32F3N3O4
Quantity 100 mg
Synonym KAD 3213 KMD 3213
Solubility Information DMSO : ≥ 50 mg/mL (100.90 mM)
SMILES O=C(N)C1=CC(C[C@@H](C)NCCOC2=CC=CC=C2OCC(F)(F)F)=CC3=C1N(CC3)CCCO
Molecular Weight (g/mol) 495.53
Formula Weight 495.53
Percent Purity 95.07%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.