Learn More
uvex Silv-Air 5110+ Premium FFP1 Flat-fold Mask
Product Code. 31910428 Shop All Uvex Products
Change view
Click to view available options
Size:
Medium
Unit Size:
Each
Product Code. Size Unit Size
31910428 Medium Each
1 options
Product Code. 31910428 Supplier uvex Supplier No. 8735111
  • ONLINE EXCLUSIVE PRICE
    51.75 €
    47.51 € / Pack of 1
    Save 4.24 €
    8% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Silv-Air premium+ product line comprises classic folding respirators that adapt to different face shapes which fits perfectly to larger face shapes.
  • The headband and integrated sealing lip in all protection classes mean the masks are easy to put on correctly as well as making them comfortable to wear, even for extended periods.
  • Individually packed
  • White colour
  • With exhalation valve
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
  • Soft sealing lip in the nose area guarantees a secure, comfortable sealed position of the mask
  • Adjustable head band offering easy handling and comfortable fit
  • Flexible and adjustable nose clip
  • Soft material edges for high wearing comfort
  • Certified FFP1 face mask to EN 149 D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • Cannot be reused
  • Respirators are not suitable for people with beards, sideburns or deep scars near the seals
TRUSTED_SUSTAINABILITY
Specifications
Color White, Blue
Design Type Flat-fold
Disposable Reusable
Filter Type FFP1
Material Polyester
Product Type Particulate Respirator
Size Medium
With Exhalation Valve Yes