Learn More
uvex Silv-Air 5320+ Premium FFP3 Flat-fold Mask
Product Code. 31910422 Shop All Uvex Products
Change view
Click to view available options
Size:
Medium
Unit Size:
Each
Product Code. Size Unit Size
31910422 Medium Each
1 options
Product Code. 31910422 Supplier uvex Supplier No. 8735321
  • ONLINE EXCLUSIVE PRICE
    118.49 €
    108.80 € / Pack of 1
    Save 9.69 €
    8% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Silv-Air premium+ product line comprises classic folding respirators that adapt to different face shapes which fits perfectly to larger face shapes.
  • The headband and integrated sealing lip in all protection classes mean the masks are easy to put on correctly as well as making them comfortable to wear, even for extended periods.
  • Individually packed
  • Grey Color
  • With exhalation valve
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
  • Soft sealing lip in the nose and chin area guarantees a secure, comfortable sealed position of the mask
  • Adjustable head band offering easy handling and comfortable fit
  • Flexible and adjustable nose clip
  • Soft material edges for high wearing comfort Certified FFP3 face mask to EN 149 D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • Additional activated carbon filter against odours, gases and vapours below threshold
  • Cannot be reused
  • Respirators are not suitable for people with beards, sideburns or deep scars near the seals
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Design Type Flat-fold
Disposable Single-use
Filter Type FFP3
Material Polyester
Product Type Particulate Respirator
Size Medium
With Exhalation Valve Yes
Latex Free Yes