Learn More
Uvex Silv-Air Classic 2200 FFP2
Product Code. 30238143 Shop All Uvex Products
Change view
Click to view available options
Quantity:
20/Pk.
Unit Size:
Pack of 20
Product Code. Quantity Unit Size
30238143 20/Pk. Pack of 20
1 options
Product Code. 30238143 Supplier Uvex Supplier No. 8762200
  • ONLINE EXCLUSIVE PRICE
    31.65 €
    28.47 € / Pack of 20
    Save 3.18 €
    10% Off
View all promotions
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Classic folding and cup-shaped respirators that fit all kinds of facial shape, with headband loops and integrated sealing lips in all protection classes.
  • Classic folding and cup-shaped respirators
  • Fit all kinds of facial shape
  • Headband loops and integrated sealing lips in all protection classes
  • Comfortable to wear for a longer period of time without becoming bothersome.
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type FFP2
Material Polypropylene, Polyester
Product Type General Purpose Particulate Respirator
Size Medium
With Exhalation Valve No
Latex Free Yes
Quantity 20/Pk.