Learn More
Uvex Silv-Air Classic 2220 FFP2 Mask
Product Code. 17955426 Shop All Uvex Products
Change view
Click to view available options
Filter Type:
FFP2
Unit Size:
Pack of 15
Product Code. Filter Type Unit Size
17955426 FFP2 Pack of 15
1 options
Product Code. 17955426 Supplier Uvex Supplier No. 8762220
  • ONLINE EXCLUSIVE PRICE
    40.60 €
    36.49 € / Pack of 15
    Save 4.11 €
    10% Off
View all promotions
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The uvex silv-Air c product range consists of both classic folding and cup-style respirators that fit a wide variety of facial shapes.
  • With headband loops and integrated sealing lips in all protection classes, it is really easy to fit the masks correctly.
  • The masks can also be worn comfortably for a longer period of time without becoming bothersome.
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type P - Oil-Proof
Color Gray
Design Type Cup/Molded
Disposable Reusable
Filter Type FFP2
Material Polypropylene, Polyester
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Medium
With Exhalation Valve Yes
Latex Free Yes
Sterility Sterile