Learn More
Uvex Silv-Air Classic 2310 Planet Mask
Product Code. 17935456 Shop All Uvex Products
Change view
Click to view available options
Filter Type:
FFP3
Unit Size:
Pack of 15
Product Code. Filter Type Unit Size
17935456 FFP3 Pack of 15
1 options
Product Code. 17935456 Supplier Uvex Supplier No. 8762313
  • ONLINE EXCLUSIVE PRICE
    74.25 €
    66.80 € / Pack of 15
    Save 7.45 €
    10% Off
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
The uvex silv-Air c product range consists of both classic folding and cup-style respirators that fit a wide variety of facial shapes.
The headband strap and integrated sealing lips in all protection classes make it easy to fit these masks correctly – and to wear them comfortably for long periods.
  • Particle-filtering preformed mask with exhalation valve for larger facial shapes
  • Valve and headband holder made of partly recycled material
  • Comfort features
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Very soft, all-round sealing lip for optimum wearer comfort and reliably tight fit
  • Exhalation valve to reduce exhalation resistance as well as heat and moisture under the mask
  • Ergonomically adapted specifically to larger facial shapes
  • FFP3 respirator compliant with EN 149:2001+A1:2009
  • D certification (mask has passed the dolomite dust test): excellent breathability even with high levels of dust
  • Not reusable
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type P - Oil-Proof
Color White
Design Type Cup/Molded
Disposable Reusable
Filter Type FFP3
Material Polypropylene, Polyester
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Medium
With Exhalation Valve Yes
Latex Free Yes
Sterility Sterile