Learn More
Uvex Silv-Air Classic 3310 FFP3 Mask
Product Code. 17945446 Shop All Uvex Products
Change view
Click to view available options
Filter Type:
FFP3
Unit Size:
Pack of 15
Product Code. Filter Type Unit Size
17945446 FFP3 Pack of 15
1 options
Product Code. 17945446 Supplier Uvex Supplier No. 8763310
  • ONLINE EXCLUSIVE PRICE
    79.54 €
    66.80 € / Pack of 15
    Save 12.74 €
    16% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The uvex silv-Air c product range consists of both classic folding and cup-style respirators that fit a wide variety of facial shapes.
  • With headband loops and integrated sealing lips in all protection classes, it is really easy to fit the masks correctly.
  • The masks can also be worn comfortably for a longer period of time without becoming bothersome.
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type P - Oil-Proof
Color White
Design Type Flat-fold
Disposable Reusable
Filter Type FFP3
Material Polypropylene, Polyester
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Medium
With Exhalation Valve Yes
Latex Free Yes
Sterility Sterile