Learn More
Uvex Silv-Air Exxcel 7312 FFP3 Mask
Product Code. 18043732 Shop All Uvex Products
Change view
Click to view available options
Filter Type:
FFP3
Unit Size:
Pack of 5
Product Code. Filter Type Unit Size
18043732 FFP3 Pack of 5
1 options
Product Code. 18043732 Supplier Uvex Supplier No. 8787312
  • ONLINE EXCLUSIVE PRICE
    32.04 €
    26.39 € / Pack of 5
    Save 5.65 €
    17% Off
View all promotions
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
All masks in the uvex silv-Air e product range feature our unique technologies to combine optimum wearer comfort with extremely low breathing resistances. This makes the products suitable for heavy work and ensures that they can be worn for long periods of time.
All masks of the uvex silv-Air e family combine our unique technologies of optimum wearer comfort with extremely low breathing resistance. The products are therefore suitable for heavy work and for being worn for long periods of time.
  • Particle-filtering preformed mask with exhalation valve
  • Soft material edges for increased wearer comfort
  • Very soft, all-around sealing lip for optimum wearer comfort and secure fit
  • Flexible, adjustable integrated nose clip
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
  • Certified FFP3 face mask to EN 149
  • D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • R label (reusable): tested for re-usability and performance, including in the second shift
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type P - Oil-Proof
Color White
Design Type Cup/Molded
Disposable Reusable
Filter Type FFP3
Material Polypropylene, Polyester
Product Type Particulate Respirator
Size Medium
With Exhalation Valve Yes
Latex Free Yes
Sterility Non-sterile