Learn More
Uvex Silv-Air Exxcel 8313 FFP3 Mask
Product Code. 18053732 Shop All Uvex Products
Change view
Click to view available options
Filter Type:
FFP3
Unit Size:
Pack of 15
Product Code. Filter Type Unit Size
18053732 FFP3 Pack of 15
1 options
Product Code. 18053732 Supplier Uvex Supplier No. 8788313
  • ONLINE EXCLUSIVE PRICE
    88.05 €
    79.21 € / Pack of 15
    Save 8.84 €
    10% Off
View all promotions
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
All masks in the uvex silv-Air e product range feature our unique technologies to combine optimum wearer comfort with extremely low breathing resistances. This makes the products suitable for heavy work and ensures that they can be worn for long periods of time.
The gray coloring on the outside of our uvex silv-Air p masks prevents them from looking dirty quickly and having to be disposed of prematurely by the user. The inside however is pure white. In addition, the 3D nose geometry for all products in this family ensures a secure fit, even for different facial shapes.
  • Particle-filtering flat-fold mask with climate-optimised exhalation valve
  • Hydrophobic material coating
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Flexible, adjustable integrated nose clip
  • Very soft, all-around sealing lip for optimal wearer comfort and secure fit
  • Individually sealed
  • Exhalation valve to reduce breathing resistance and minimise heat and moisture build-up inside the mask
  • FFP3 respirator certified in accordance with EN 149:2001+A1:2009
  • D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type P - Oil-Proof
Color White
Design Type Flat-fold
Disposable Reusable
Filter Type FFP3
Material Polypropylene, Polyester
Product Type Particulate Respirator
Size Medium
With Exhalation Valve Yes
Latex Free Yes
Sterility Non-sterile