Learn More
Uvex Silv-Air Premium 5310 FFP3
Product Code. 30238145 Shop All Uvex Products
Change view
Click to view available options
Quantity:
15/Pk.
Unit Size:
Pack of 15
Product Code. Quantity Unit Size
30238145 15/Pk. Pack of 15
1 options
Product Code. 30238145 Supplier Uvex Supplier No. 8735310
  • ONLINE EXCLUSIVE PRICE
    116.00 €
    113.24 € / Pack of 15
    Save 2.76 €
    2% Off
View all promotions
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Classic, white flat-fold respirators that fit all kinds of facial shape, with headband loops and integrated sealing lips in all protection classes.
  • Classic, white flat-fold respirators
  • Fit all kinds of facial shape
  • Headband loops and integrated sealing lips in all protection classes
  • Comfortable to wear for a longer period of time without becoming bothersome
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Color White
Design Type Flat-fold
Disposable Limited-use
Filter Type FFP3
Material Polypropylene, Polyester
Product Type General Purpose Particulate Respirator
Size Small
With Exhalation Valve Yes
Latex Free No
Quantity 15/Pk.