Learn More
Silver diethyldithiocarbamate, ACS, Thermo Scientific™
POA
Chemical Identifiers
| CAS | 1470-61-7 |
|---|---|
| Molecular Formula | C5H10AgNS2 |
| Molecular Weight (g/mol) | 256.13 |
| MDL Number | MFCD00004929 |
| InChI Key | NSVHDIYWJVLAGH-UHFFFAOYSA-M |
| Synonym | silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate |
| PubChem CID | 3034078 |
| IUPAC Name | silver;N,N-diethylcarbamodithioate |
| SMILES | CCN(CC)C(=S)[S-].[Ag+] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C5H10AgNS2 | |
| MFCD00004929 | |
| silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate | |
| silver;N,N-diethylcarbamodithioate |
Specifications
| 1470-61-7 | |
| MFCD00004929 | |
| It is soluble in water and pyridine. | |
| CCN(CC)C(=S)[S-].[Ag+] | |
| 256.13 | |
| 256.14 | |
| Odorless | |
| Yellow to pale brown powder |
| C5H10AgNS2 | |
| silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate | |
| NSVHDIYWJVLAGH-UHFFFAOYSA-M | |
| silver;N,N-diethylcarbamodithioate | |
| 3034078 | |
| ACS | |
| Light sensitive | |
| Silver diethyldithiocarbamate, For analysis ACS |
Sicurezza e movimentazione
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 216-003-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.