missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfate, pure, 10 g/l solution in sulfuric acid

Product Code. 10237460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10237460 1 L 1 L
10247460 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10237460 Supplier Thermo Scientific Acros Supplier No. 279480010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver sulfate
Color Colorless
Name Note 10g/L Solution in Sulfuric Acid
CAS 10294-26-5, 7664-93-9
Molecular Formula Ag2H2O4S
Linear Formula Ag2SO4
MDL Number MFCD00003407
Fieser 01,1015; 03,254; 04,435
Density 1.8400g/mL
Merck Index 15, 8668
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES [Ag+].[Ag+].OS(O)(=O)=O
IUPAC Name disilver(1+) sulfuric acid
Molecular Weight (g/mol) 313.81
PubChem CID 159865
Formula Weight 311.79
Grade Pure
Packaging Glass bottle
Specific Gravity 1.84
Quantity 1 L
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Silver sulfate, 10 g/l solution in sulfuric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.