missing translation for 'onlineSavingsMsg'
Learn More

Silver Sulfate Solution 5% (w/v) (d=1.84) In Sulfuric Acid, for COD, Fisher Chemical

Product Code. 10030230 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10030230 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10030230 Supplier Fisher Chemical Supplier No. J/7410/PB17
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 10294-26-5
Molecular Formula Ag2H2O4S
MDL Number 3407
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES [Ag+].[Ag+].OS(O)(=O)=O
IUPAC Name disilver(1+) sulfuric acid
Molecular Weight (g/mol) 313.81
PubChem CID 159865
Formula Weight 311.79g/mol
Packaging Plastic Coated Glass bottle
pH 1
Quantity 2.5 L
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Silver sulfate, 10 g/l solution in sulfuric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.