missing translation for 'onlineSavingsMsg'
Learn More

Simazine, Dr. Ehrenstorfer

Product Code. 30316152
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316152 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316152 Supplier Dr. Ehrenstorfer Supplier No. DREC16950000100MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Simazine
CAS 122-34-9
Analyte or Component Names Simazine
Shipping Condition +20°C
Molecular Formula C7 H12 Cl N5
Certifications/Compliance ISO 17034
Synonym 1,3,5-Triazine-2,4-diamine, 6-chloro-N2,N4-diethyl-, 1,3,5-Triazine-2,4-diamine, 6-chloro-N,N'-diethyl- (9CI), s-Triazine, 2-chloro-4,6-bis(ethylamino)- (8CI), 6-Chloro-N2,N4-diethyl-1,3,5-triazine-2,4-diamine, 2,4-Bis(ethylamino)-6-chloro-s-triazine, 2,4-Di(ethylamino)-6-chloro-1,3,5-triazine, 2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine, 2-Chloro-4,6-bis(ethylamino)-s-triazine, 2-Chloro-4,6-bis(ethylamino)-sym-triazine, 2-Chloro-4,6-di(ethylamino)triazine, 4,6-Bis(ethylamino)-2-chlorotriazine, 6-Chloro-N2,N4-diethyl-1,3,5-triazine-2,4-diamine, Aktinit S, Aquazine, Azotop, Batazina FLO, Bitemol S 50, CAT, CAT (pesticide), CDT, CET, G 27692, Geigy 27692, Gesatop, Gesatop 50, Gesatop D, H 1803, Herbatoxol S, Herbazin, Herbazin 50, Herbex, Herbex (pesticide), Herboxy, Hungazin DT, NSC 25999, Premazine, Princep, Radocon, Radokor, Sim-Trol 4L, Simanex, Simatox, Simatsin-neste, Simazin, Simazin S 50, Simazine 50, Simazine 80W, Symazine, Tafazine, Tafazine 50W, Taphazine, Triazine A 384, W 6658, Yrodazin
InChI Formula InChI=1S/C7H12ClN5/c1-3-9-6-11-5(8)12-7(13-6)10-4-2/h3-4H2,1-2H3,(H2,9,10,11,12,13)
SMILES CCNc1nc(Cl)nc(NCC)n1
IUPAC Name 6-chloro-2-N,4-N-diethyl-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol) 201.66
Quantity 100 mg
Formula Weight 201.0781 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.