missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simian Immunodeficiency Virus gp120 Antibody [Janelia Fluor™ 549] - (isolate F236), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43772JF549
This item is not returnable.
View return policy
Description
Simian Immunodeficiency Virus gp120 Polyclonal antibody specifically detects Simian Immunodeficiency Virus gp120 in Virus samples. It is validated for Western Blot
Specifications
| Simian Immunodeficiency Virus gp120 | |
| Polyclonal | |
| Western Blot | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot | |
| Janelia Fluor 549 | |
| 50 mM Sodium Borate | |
| Produced in rabbits immunized with purified, recombinant Simian Immunodeficiency Virus gp120 (isolate F236) (RefSeq#: CAA32487.1; Gln24-Arg531) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction