missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Performance Plastics Single-Use Bioprocess Bags, Standard 2D, 2 ports
Click to view available options
Capacity (Metric):
50 mL
150 mL
250 mL
500 mL
Tubing Length Line 1:
12 in.
Description
- Available in sizes from 50 mL to 500 mL.
- Constructed with LLDPE as the fluid contact layer and EVOH as the gas barrier, ensuring chemical compatibility, durability, and protection against oxygen ingress.
- Designed with top or bottom drain configurations to meet diverse process requirements and maximize ease of use.
- Needle-free sampling ports.
- Small volume bags feature integrated hanging capability and handles.
- Sealable, weldable tubing on standard designs ensures leak-free connections and compatibility with automated welding equipment.
- Available in both standard and custom designs.
- Available as manifold systems.
- Ideal for cell culture media, buffer preparation, product storage, and sampling across upstream and downstream bioprocessing operations.
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Dimensions | 5.22 x 3.85 in. |
| Film Description | Saint-Gobain 5-layer film |
| Port Detail | 2 Ports |
| Tubing Length Line 1 | 12 in. |
| Tubing Length Line 2 | 12 in. |
| Tubing Material Line 1 | C-Flex 374 |
| Tubing Material Line 2 | C-Flex 374 |
| Tubing Style | C-Flex |
| Product Type | Standard 2D Bioprocess Bag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction