missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Performance Plastics Single-Use Bioprocess Bags, Standard 2D, 3 ports

Product Code. 31723090
Change view
Click to view available options
Capacity (Metric):
50 mL
150 mL
250 mL
500 mL
1 L
2 L
3 L
5 L
10 L
20 L
Tubing Length Line 1:
6 in.
20 in.
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Tubing Length Line 1
31723090 1 L 6 in.
31723072 1 L 20 in.
31723088 2 L 6 in.
31723085 2 L 20 in.
31723080 3 L 6 in.
31723067 3 L 20 in.
31723083 5 L 6 in.
31723089 5 L 20 in.
31723082 10 L 6 in.
31723061 10 L 20 in.
31723081 20 L 6 in.
31723075 20 L 20 in.
31723068 50 mL 6 in.
31723077 150 mL 6 in.
31723076 150 mL 20 in.
31723087 250 mL 6 in.
31723073 250 mL 20 in.
31723084 500 mL 6 in.
31723065 500 mL 20 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Product Code. 31723090 Supplier Saint-Gobain Performance Plastics Supplier No. BP0001L
  • ONLINE EXCLUSIVE PRICE
    1432.43 €
    1128.93 € / Pack of 20
    Save 303.50 €
    21% Off
 Restricted Item
Estimated Shipment: 27-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Built with multilayer films for strong gas barrier protection ensuring cGMP-compliant and animal-free performance

  • Available in sizes from 50 mL to 20 L.
  • Constructed with LLDPE as the fluid contact layer and EVOH as the gas barrier, ensuring chemical compatibility, durability, and protection against oxygen ingress.
  • Designed with top or bottom drain configurations to meet diverse process requirements and maximize ease of use.
  • Needle-free sampling ports.
  • Small volume bags feature integrated hanging capability and handles.
  • Sealable, weldable tubing on standard designs ensures leak-free connections and compatibility with automated welding equipment.
  • Available in both standard and custom designs.
  • Available as manifold systems.
  • Ideal for cell culture media, buffer preparation, product storage, and sampling across upstream and downstream bioprocessing operations.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 L
Dimensions 13.50 x 6.48 in.
Film Description Saint-Gobain 5-layer film
Port Detail 3 Ports
Tubing Length Line 1 6 in.
Tubing Length Line 3 6 in.
Tubing Length Line 2 6 in.
Tubing Material Line 3 C-Flex 374
Tubing Material Line 1 C-Flex 374
Tubing Material Line 2 C-Flex 374
Tubing Style C-Flex
Product Type Standard 2D Bioprocess Bag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.