missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly

Product Code. 16679561 Shop All Masterflex Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
2 L
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
16679561 125 mL Each
16689561 250 mL Each
16699561 500 mL Each
16609571 1 L Each
16619571 2 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16679561 Supplier Masterflex™ Supplier No. 9564420
Estimated Shipment: 12-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly safely transfers, samples, and stores biopharmaceutical products and reagents

  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
  • Assemblies are gamma irradiated - eliminates labor and validation costs associated with assembly and cleaning

Specifications

Description Gamma Irradiated
Capacity (Metric) 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.