Learn More
Honeywell™ Single-Use Molded Facemask, FFP2 NRD with Valve
Product Code. 15866825
Change view
Click to view available options
:
Size: Medium
Product Code.
15866825 Size: Medium
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15866825 Supplier Honeywell™ Supplier No. 1007224
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Rigid shell facemask for protection from particulates even in moistened and heavy loading environments.
  • Rigid shell: shape maintained even in moistened and heavy loading environments
  • Soft nose seal with absorption properties and a tight sealing film for comfort and fit
  • Preformed and colored nosebridge to save time to perfectly adjust the mask and to instantly identify its protection level
  • Large internal volume to easily communicate and breathe
  • Shielded exhalation valve with an exclusive design for improved protection and breathing comfort
  • Twin elastic straps, ultra flexible and strong, stapled outside the filtering area to avoid any risk of leakages
  • Lightweight (15 g) for better wearer acceptance
  • Free of latex, silicone, and PVC
TRUSTED_SUSTAINABILITY
Specifications
Design Type Cup/Molded
Size Medium
With Exhalation Valve Yes
For Use With (Application) Agriculture, automotive, building, construction, foundry, iron/steel, mining, quarrying, textile, wood industry
Includes Double elastics
Latex Free Yes
Weight (Metric) 15 g