missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIPA1L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06158-100ul
This item is not returnable.
View return policy
Description
SIPA1L1 Polyclonal antibody specifically detects SIPA1L1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SIPA1L1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| E6-targeted protein 1, FLJ37213, H_DJ1140G11.1, high-risk human papilloma viruses E6 oncoproteins targeted protein 1, KIAA0440, signal-induced proliferation-associated 1 like 1 | |
| Produced in rabbits immunized with E. coli-derived Human SIPA1L1 fragment. | |
| 100 μg | |
| Cancer | |
| 26037 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction