missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF11 Monoclonal Antibody (224812)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561015
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Probably involved in nervous system development and function.
Specifications
| FGF11 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q92914 | |
| FGF11 | |
| E. coli-derived recombinant human FGF-11, Met1-Pro225 (predicted). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 224812 | |
| Unconjugated | |
| FGF11 | |
| FGF; Fgf11; Fgf-11; Fhf3; FHF-3; Fibroblast growth factor; Fibroblast growth factor 11; fibroblast growth factor homologous factor 3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2256 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction