missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPP9 Monoclonal Antibody (988515)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561551
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a protein that is a member of the S9B family in clan SC of the serine proteases. The protein has been shown to have post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Although the activity of this protein is similar to that of dipeptidyl peptidase 4 (DPP4), it does not appear to be membrane bound. In general, dipeptidyl peptidases appear to be involved in the regulation of the activity of their substrates and have been linked to a variety of diseases including type 2 diabetes, obesity and cancer. Several transcript variants of this gene have been described but not fully characterized.
Specifications
| DPP9 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q86TI2 | |
| DPP9 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human DPP9, Arg2-Leu892. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| 988515 | |
| Unconjugated | |
| DPP9 | |
| 6430584G11Rik; A330078I11; Dipeptidyl peptidase 9; dipeptidyl peptidase IV-related protein 2; dipeptidyl peptidase IV-related protein-2; dipeptidyl peptidase IX; Dipeptidyl peptidase-like protein 9; dipeptidylpeptidase 9; dipeptidyl-peptidase 9; DKFZp762F117; DP9; DPLP9; DPP IX; DPP9; DPRP2; DPRP-2; FLJ16073 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 91039 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction