missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRP beta/gamma Polyclonal Antibody

Description
Reconstitute at 0.2 mg/mL in sterile PBS.
SIRP beta and SIRP gamma are two members of the signal regulatory protein (SIRP) family. These proteins are transmembrane receptors that are expressed on the surface of various cells, including immune cells and neurons. The function of SIRP beta and SIRP gamma is to regulate cellular signaling pathways that are involved in immune responses, cell adhesion, and phagocytosis. Specifically, these proteins interact with the protein tyrosine phosphatase SHP-1, which is a negative regulator of immune cell activation. By binding to SHP-1, SIRP beta and SIRP gamma can inhibit immune cell activation and promote immune tolerance. In addition to their role in immune regulation, SIRP beta and SIRP gamma have also been implicated in neuronal signaling and synaptic plasticity. These proteins are thought to play a role in the formation and maintenance of synapses, as well as in the regulation of neuronal activity.
Specifications
Specifications
| Antigen | SIRP beta/gamma |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | SIRPB1 |
| Gene Accession No. | O00241, Q9P1W8 |
| Gene Symbols | SIRPB1, SIRPG |
| Host Species | Goat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?