missing translation for 'onlineSavingsMsg'
Learn More

Six elements precious metals - Au, Ir, Pd, Pt, Rh, Ru for ICP 100mg/L HNO3 matrix, Fisher chemical™

Product Code. 12996183 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12996183 500 mL 500mL
12976183 50 mL 50mL
12986183 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12996183 Supplier Fisher Chemical Supplier No. MS 004 500
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Chemical Name or Material Six Elements (Precious Metals)
Packaging Bottle
Analyte or Component Names Au,Ir,Pd,Pt,Rh,Ru
Physical Form Liquid
Concentration 100 mg/L
Type ICP Multi-element Standard Solution
Element Au, Ir, Pd, Pt, Rh, Ru
Solvent or Matrix 10% HCl
Quantity 500 mL
compliance-icons
Product Identifier
  • Multi-element Standard Solution for ICP. 6 Components in HCl 10% (MS 004)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.